AMBH2D70260E Ambeed, Inc.

4,4,5,5-Tetramethyl-2-(4-(trifluoromethyl)benzyl)-1,3,2-dioxaborolane

  • MDL number MFCD10698521
  • Cas Number : 475250-46-5
  • Molecular Weight : 286.1000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/475250-46-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/475250-46-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart