AMBH2D70263D Ambeed, Inc.

4-Vinylidenebis(N,N-dimethylaniline)

  • MDL number MFCD00192575
  • Cas Number : 7478-69-5
  • Molecular Weight : 266.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/7478-69-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7478-69-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart