AMBH2D7026CA Ambeed, Inc.

Ethyl 4-amino-2-nitrobenzoate

  • MDL number MFCD06656140
  • Cas Number : 84228-46-6
  • Molecular Weight : 210.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 397.6�0�t 760 mmHg
Iupac Name ethyl 4-amino-2-nitrobenzoate
Inchi Code 1S/C9H10N2O4/c1-2-15-9(12)7-4-3-6(10)5-8(7)11(13)14/h3-5H,2,10H2,1H3
Inchi Key YOMQIQZTFBDFSR-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 194.3
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 397.6�0�t 760 mmHg
Product Link https://www.ambeed.com/products/84228-46-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/84228-46-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart