AMBH2D7026CD Ambeed, Inc.

4-Bromo-5-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD00464256
  • Cas Number : 84547-93-3
  • Molecular Weight : 235.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-202
Synonym N/A
Purity 95
Flash Point 227.3
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 452.3�0�t 760 mmHg
Product Link https://www.ambeed.com/products/84547-93-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/84547-93-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart