AMBH2D7026E5 Ambeed, Inc.

Metoclopramide hydrochloride hydrate

  • MDL number MFCD00150634
  • Cas Number : 54143-57-6
  • Molecular Weight : 336.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/54143-57-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/54143-57-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart