AMBH2D702726 Ambeed, Inc.

N-Boc-2-Morpholinecarbaldehyde

  • MDL number MFCD06410632
  • Cas Number : 218594-02-6
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-butyl 2-formylmorpholine-4-carboxylate
Purity 97
Flash Point 140.7°C
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form Solid or liquid
Shipping Temperature Cold-chain transportation
Country of Origin China
Boiling point 309.1°C at 760 mmHg
Product Link https://www.ambeed.com/products/218594-02-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/218594-02-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart