AMBH2D702750 Ambeed, Inc.

4-Bromo-2-ethoxypyridine

  • MDL number MFCD00234310
  • Cas Number : 57883-26-8
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 5.5-7
Purity 98
Flash Point 87.6
Storage temp. Inert atmosphere,2-8
Physical Form Liquid or Solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 221.2�t 760 mmHg
Product Link https://www.ambeed.com/products/57883-26-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/57883-26-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart