AMBH2D702752 Ambeed, Inc.

4-Bromo-1-(dimethoxymethyl)-2-fluorobenzene

  • MDL number MFCD07780632
  • Cas Number : 439814-87-6
  • Molecular Weight : 249.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/439814-87-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/439814-87-6.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart