AMBH2D70283E Ambeed, Inc.

4'-Dodecyl-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD08062573
  • Cas Number : 57125-49-2
  • Molecular Weight : 347.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Flash Point 248.7
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 484.4�t 760 mmHg
Product Link https://www.ambeed.com/products/57125-49-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/57125-49-2.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart