AMBH2D702870 Ambeed, Inc.

4-Fluoro-3-formylbenzoic acid

  • MDL number MFCD06411555
  • Cas Number : 845885-90-7
  • Molecular Weight : 168.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/845885-90-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/845885-90-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart