AMBH2D70289B Ambeed, Inc.

2-Amino-3H-pyrrolo[3,2-d]pyrimidin-4(5H)-one

  • MDL number MFCD09540493
  • Cas Number : 65996-58-9
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/65996-58-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/65996-58-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart