AMBH2D702921 Ambeed, Inc.

4-methoxypicolinonitrile

  • MDL number MFCD08062857
  • Cas Number : 36057-44-0
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-121
Purity 97
Flash Point 110
Storage temp. Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 257.9�t 760 mmHg
Product Link https://www.ambeed.com/products/36057-44-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/36057-44-0.html
pictograms GHS07
hstatements H302+H312+H332-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart