AMBH2D702D26 Ambeed, Inc.

4-Methylpyridazin-3(2H)-one

  • MDL number MFCD08460985
  • Cas Number : 33471-40-8
  • Molecular Weight : 110.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-153
Synonym 4-Methylpyridazin-3-ol
Purity 97
Storage temp. Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 358.1�t 760 mmHg
Product Link https://www.ambeed.com/products/33471-40-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/33471-40-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart