AMBH2D702D47 Ambeed, Inc.

2-Oxo-2-(p-tolyl)acetaldehyde hydrate

  • MDL number MFCD00235014
  • Cas Number : 7466-72-0
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Store in freezer, under -20°C
Physical Form solid
Shipping Temperature Shipped in cold pack
Country of Origin China
Product Link https://www.ambeed.com/products/7466-72-0.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/7466-72-0.html
pictograms GHS07
hstatements H302-H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart