AMBH303C44E3 Ambeed, Inc.

1-Chloro-3-iodo-5-nitrobenzene

  • MDL number MFCD13194575
  • Cas Number : 123158-76-9
  • Molecular Weight : 283.4500

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Physical Form solid
Shipping Temperature Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/123158-76-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/123158-76-9.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart