AMBH303C44EA Ambeed, Inc.

B-Raf inhibitor 1

  • MDL number MFCD22571725
  • Cas Number : 1093100-40-3
  • Molecular Weight : 478.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1093100-40-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1093100-40-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart