AMBH303C471B Ambeed, Inc.

1-(Furan-2-ylmethyl)pyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD08543791
  • Cas Number : 953409-03-5
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/953409-03-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/953409-03-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart