AMBH303C4944 Ambeed, Inc.

1-(6-((Benzyloxy)bis(4-methoxyphenyl)methyl)-4-(2-((tert-butyldimethylsilyl)oxy)ethoxy)-5-hydroxytetrahydro-2H-pyran-3-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

  • MDL number N/A
  • Cas Number : 1956306-32-3
  • Molecular Weight : 732.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1956306-32-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1956306-32-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart