AMBH303C496C Ambeed, Inc.

Trimethobenzamide HCl

  • MDL number MFCD00057999
  • Cas Number : 554-92-7
  • Molecular Weight : 424.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/554-92-7.html

Safety Information

signalword Danger
pcodes P261-P280-P284-P301+P312+P330-P304+P340-P342+P311
MSDS https://www.ambeed.com/products/554-92-7.html
pictograms GHS07,GHS08
hstatements H302-H317-H334
Availability details for
Quantity
  • Availability
  • Add to Cart