AMBH303C49B0 Ambeed, Inc.

Hexadecyl 3-amino-4-chlorobenzoate

  • MDL number MFCD00186289
  • Cas Number : 143269-74-3
  • Molecular Weight : 396.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/143269-74-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/143269-74-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart