AMBH303C49C9 Ambeed, Inc.

2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl

  • MDL number MFCD11045061
  • Cas Number : 95759-51-6
  • Molecular Weight : 360.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/95759-51-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/95759-51-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart