AMBH303C4A04 Ambeed, Inc.

Ethyl 3-methylpiperidine-3-carboxylate hydrochloride

  • MDL number MFCD15142861
  • Cas Number : 176523-95-8
  • Molecular Weight : 207.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/176523-95-8.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/176523-95-8.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart