AMBH303C4A0D Ambeed, Inc.

Phenyl (4-(4-(4-hydroxyphenyl)piperazin-1-yl)phenyl)carbamate

  • MDL number MFCD03784249
  • Cas Number : 184177-81-9
  • Molecular Weight : 389.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/184177-81-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/184177-81-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart