AMBH303C4A28 Ambeed, Inc.

JSH-23

  • MDL number MFCD09753595
  • Cas Number : 749886-87-1
  • Molecular Weight : 240.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/749886-87-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/749886-87-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart