AMBH303C4A3C Ambeed, Inc.

1,1,4,4,6-Pentamethyl-7-(1-(p-tolyl)vinyl)-1,2,3,4-tetrahydronaphthalene

  • MDL number MFCD28369474
  • Cas Number : 1688656-72-5
  • Molecular Weight : 318.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1688656-72-5.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1688656-72-5.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart