AMBH303C4A7D Ambeed, Inc.

(4-Fluoro-3-isopropylphenyl)boronic acid

  • MDL number MFCD30489011
  • Molecular Weight : 182.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/P000083034.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/P000083034.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart