AMBH303C4A81 Ambeed, Inc.

Methyl 2-(4-bromobenzoyl)-3-(methylamino)but-2-enoate

  • MDL number MFCD30729997
  • Cas Number : 1423699-80-2
  • Molecular Weight : 312.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1423699-80-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1423699-80-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart