AMBH303C4A8E Ambeed, Inc.

2,4,6-Tris(3-bromophenyl)-1,3,5-triazine

  • MDL number MFCD28964709
  • Cas Number : 890148-78-4
  • Molecular Weight : 546.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/890148-78-4.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/890148-78-4.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart