AMBH303C4ADE Ambeed, Inc.

Ac-His-Trp-Ala-Val-Gly-His-Leu-NH2

  • MDL number MFCD00237057
  • Cas Number : 121432-21-1
  • Molecular Weight : 944.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 1478.0�0�t 760 mmHg
Iupac Name (2S,8S,11S,14S,17S)-2,17-bis((1H-imidazol-4-yl)methyl)-14-((1H-indol-3-yl)methyl)-3,6,9,12,15,18-hexaamino-N-((S)-1-amino-4-methyl-1-oxopentan-2-yl)-8-isopropyl-11-methyl-4,7,10,13,16,19-hexaoxoicosanamide
Inchi Code 1S/C47H69N13O8/c1-21(2)11-34(46(54)67)60-47(68)31(14-27-18-56-20-59-27)39(51)35(62)15-32(48)45(66)36(22(3)4)41(53)42(63)23(5)37(49)43(64)29(12-25-16-57-33-10-8-7-9-28(25)33)40(52)44(65)30(38(50)24(6)61)13-26-17-55-19-58-26/h7-10,16-23,29-32,34,36-41,57H,11-15,48-53H2,1-6H3,(H2,54,67)(H,55,58)(H,56,59)(H,60,68)/t23-,29-,30-,31-,32?,34-,36-,37?,38?,39?,40?,41?/m0/s1
Inchi Key LZCZGBCZVOUXDC-VIZOZCGASA-N

Description

Synonym (S)-2-((2S,8S,11S,14S,17S)-2,17-Bis((1H-imidazol-4-yl)methyl)-14-((1H-indol-3-yl)methyl)-8-isopropyl-11-methyl-4,7,10,13,16,19-hexaoxo-3,6,9,12,15,18-hexaazaicosanamido)-4-methylpentanamide
Purity 97%
Flash Point 847.7
Storage temp. Sealed in dry,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 1478.0�0�t 760 mmHg
Product Link https://www.ambeed.com/products/121432-21-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/121432-21-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart