AMBH303C4B12 Ambeed, Inc.

7-Methoxy-2-oxo-1,2,3,4-tetrahydroquinoline-4-carbonitrile

  • MDL number MFCD27996313
  • Cas Number : 1567360-56-8
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1567360-56-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1567360-56-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart