AMBH303C4B20 Ambeed, Inc.

7-Benzyl-1-oxo-2,7-diazaspiro[3.5]nonane

  • MDL number MFCD27987069
  • Cas Number : 1334536-88-7
  • Molecular Weight : 230.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1334536-88-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1334536-88-7.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart