AMBH303C4B61 Ambeed, Inc.

2-(4-Methylpiperazin-1-ylmethyl)benzylamine

  • MDL number MFCD06797802
  • Cas Number : 879896-50-1
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/879896-50-1.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/879896-50-1.html
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart