AMBH303C4B64 Ambeed, Inc.

3-(4-Methylphenyl)-3-oxetanamine hydrochloride

  • MDL number MFCD19381737
  • Cas Number : 1322200-77-0
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1322200-77-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1322200-77-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart