AMBH303C4BA9 Ambeed, Inc.

2-Amino-5-bromo-3-piperidin-1-ylpyrazine

  • MDL number MFCD08460061
  • Cas Number : 90674-84-3
  • Molecular Weight : 257.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/90674-84-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/90674-84-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart