AMBH303C4BF0 Ambeed, Inc.

AS-1842856

  • MDL number MFCD30718173
  • Cas Number : 836620-48-5
  • Molecular Weight : 347.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/836620-48-5.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/836620-48-5.html
pictograms GHS07,GHS09
hstatements H302-H410
Availability details for
Quantity
  • Availability
  • Add to Cart