AMBH303C4C31 Ambeed, Inc.

4-Cyano-3-fluorophenyl 4-(trans-4-butylcyclohexyl)benzoate

  • MDL number MFCD11053366
  • Cas Number : 92118-83-7
  • Molecular Weight : 379.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/92118-83-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/92118-83-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart