AMBH303C4C33 Ambeed, Inc.

8-Benzyl 1,3-di-tert-butyl 2,4-dioxo-1,3,8-triazaspiro[4.5]decane-1,3,8-tricarboxylate

  • MDL number MFCD29037425
  • Cas Number : 1005187-65-4
  • Molecular Weight : 503.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1005187-65-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1005187-65-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart