AMBH303C4C43 Ambeed, Inc.

Benzofuran-6-ol

  • MDL number MFCD08669481
  • Cas Number : 13196-11-7
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108°C
Purity 95
Storage temp. Sealed in dry,Room Temperature
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/13196-11-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/13196-11-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart