AMBH303C4C4A Ambeed, Inc.

5-Chloro-2-(difluoromethoxy)pyridine

  • MDL number MFCD13185854
  • Cas Number : 1214323-40-6
  • Molecular Weight : 179.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1214323-40-6.html

Safety Information

signalword Warning
pcodes P264-P270-P301+P312-P330
MSDS https://www.ambeed.com/products/1214323-40-6.html
pictograms GHS07
hstatements H302-H315-H320-H335
Availability details for
Quantity
  • Availability
  • Add to Cart