AMBH303C4C53 Ambeed, Inc.

1,3-Bis(dodecyloxy)benzene

  • MDL number N/A
  • Cas Number : 41662-92-4
  • Molecular Weight : 446.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/41662-92-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/41662-92-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart