AMBH303C4C63 Ambeed, Inc.

tert-Butyl 3-(methylamino)piperidine-1-carboxylate hydrochloride

  • MDL number MFCD09878919
  • Cas Number : 1187927-08-7
  • Molecular Weight : 250.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1187927-08-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1187927-08-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart