AMBH303C4C94 Ambeed, Inc.

(Z)-N-(1-(4-Bromophenyl)-1-chloro-3-oxo-3-(piperidin-1-yl)prop-1-en-2-yl)benzamide

  • MDL number MFCD29037406
  • Cas Number : 1800044-79-4
  • Molecular Weight : 447.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1800044-79-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1800044-79-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart