AMBH303C4C98 Ambeed, Inc.

7-Methoxy-4-methylcoumarin

  • MDL number MFCD00009773
  • Cas Number : 2555-28-4
  • Molecular Weight : 190.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/2555-28-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2555-28-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart