AMBH303C4CA3 Ambeed, Inc.

1-(2-Fluorophenyl)propan-1-amine hydrochloride

  • MDL number MFCD28126467
  • Cas Number : 1955554-65-0
  • Molecular Weight : 189.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1955554-65-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1955554-65-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart