AMBH303C4CCB Ambeed, Inc.

Ethyl 2-(6-bromoimidazo[1,2-a]pyridin-3-yl)acetate

  • MDL number MFCD09839780
  • Cas Number : 603311-76-8
  • Molecular Weight : 283.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/603311-76-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/603311-76-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart