AMBH303C4CCC Ambeed, Inc.

7-Bromoquinolin-5-ol

  • MDL number MFCD11878500
  • Cas Number : 1261677-80-8
  • Molecular Weight : 224.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1261677-80-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1261677-80-8.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart