AMBH303C4D1A Ambeed, Inc.

2-Amino-5-nitrobenzenesulfonic acid

  • MDL number MFCD00035755
  • Cas Number : 96-75-3
  • Molecular Weight : 218.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/96-75-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/96-75-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart