AMBH303C4D2B Ambeed, Inc.

4-Phenyl-2,8-diazaspiro[4.5]decan-3-one hydrochloride

  • MDL number MFCD30489144
  • Cas Number : 2044705-27-1
  • Molecular Weight : 266.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/2044705-27-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/2044705-27-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart