AMBH303C4D67 Ambeed, Inc.

NSC 66811

  • MDL number MFCD08823776
  • Cas Number : 6964-62-1
  • Molecular Weight : 340.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 529.5�0�t 760 mmHg
Iupac Name 7-[anilino(phenyl)methyl]-2-methyl-8-quinolinol
Inchi Code 1S/C23H20N2O/c1-16-12-13-18-14-15-20(23(26)22(18)24-16)21(17-8-4-2-5-9-17)25-19-10-6-3-7-11-19/h2-15,21,25-26H,1H3
Inchi Key WEENRMPCSWFMTE-UHFFFAOYSA-N

Description

Purity 95%
Flash Point 274
Storage temp. Keep in dark place,Inert atmosphere,2-8
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 529.5�0�t 760 mmHg
Product Link https://www.ambeed.com/products/6964-62-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6964-62-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart