AMBH303C4D80 Ambeed, Inc.

2-Methylimidazo[1,2-a]pyridin-8-ylamine hydrochloride

  • MDL number MFCD07367043
  • Cas Number : 173159-45-0
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/173159-45-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/173159-45-0.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart